Assay Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024188.6 | NP_077150.1 | 5 - 6 | 903 | 102 | |
| GenBank mRNA | AB085609.1 | - | 5 - 6 | 645 | 102 | |
| AK009027.1 | - | 5 - 6 | 683 | 102 | ||
| AK010790.1 | - | 5 - 6 | 646 | 102 | ||
| AK011354.1 | - | 5 - 6 | 583 | 102 | ||
| AK078086.1 | - | 5 - 6 | 903 | 102 | ||
| AK146158.1 | - | 5 - 6 | 102 | |||
| AK146342.1 | - | 5 - 6 | 648 | 102 | ||
| AK151662.1 | - | 5 - 6 | 642 | 102 | ||
| AK151695.1 | - | 5 - 6 | 633 | 102 | ||
| AK167681.1 | - | 5 - 6 | 733 | 102 | ||
| AK181314.1 | - | 1 - 2 | 118 | 102 | ||
| AK184597.1 | - | 1 - 2 | 119 | 102 | ||
| BC003422.1 | - | 5 - 6 | 607 | 2598824 | 102 | |
| EF095212.1 | - | 2 - 3 | 296 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oxct1
Genomic Map