Assay Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024188.6 | NP_077150.1 | 8 - 9 | 1183 | 77 | |
| GenBank mRNA | AB085609.1 | - | 8 - 9 | 925 | 77 | |
| AK009027.1 | - | 8 - 9 | 963 | 77 | ||
| AK011354.1 | - | 8 - 9 | 863 | 77 | ||
| AK078086.1 | - | 8 - 9 | 1183 | 77 | ||
| AK146158.1 | - | 8 - 9 | 941 | 77 | ||
| AK146342.1 | - | 8 - 9 | 928 | 77 | ||
| AK151662.1 | - | 8 - 9 | 922 | 77 | ||
| AK151695.1 | - | 8 - 9 | 913 | 77 | ||
| AK167681.1 | - | 8 - 9 | 1013 | 77 | ||
| BC003422.1 | - | 8 - 9 | 887 | 2598824 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oxct1
Genomic Map