Assay Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024188.6 | NP_077150.1 | 14 - 15 | 1683 | 97 | |
| GenBank mRNA | AB085609.1 | - | 14 - 15 | 1425 | 97 | |
| AK010029.1 | - | 1 - 2 | 66 | 97 | ||
| AK011354.1 | - | 14 - 15 | 1363 | 97 | ||
| AK078086.1 | - | 14 - 15 | 1681 | 97 | ||
| AK146158.1 | - | 14 - 15 | 1441 | 97 | ||
| AK146342.1 | - | 14 - 15 | 1428 | 97 | ||
| AK151662.1 | - | 14 - 15 | 1422 | 97 | ||
| AK151695.1 | - | 14 - 15 | 1413 | 97 | ||
| AK167681.1 | - | 14 - 15 | 1513 | 97 | ||
| AK185909.1 | - | 4 - 5 | 258 | 97 | ||
| BC003422.1 | - | 14 - 15 | 1387 | 2598824 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oxct1
Genomic Map