Assay Details
| Gene Symbol: Timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024273.2 | NP_077235.2 | 4 - 5 | 646 | 73 | |
| GenBank mRNA | AK081781.1 | - | 4 - 5 | 589 | 73 | |
| BC002243.1 | - | 4 - 5 | 608 | 3492342 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Timmdc1
Genomic Map