Assay Details
| Gene Symbol: Lix1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025681.2 | NP_079957.2 | 1 - 2 | 342 | 71 | |
| GenBank mRNA | AF351204.1 | - | 1 - 2 | 83 | 71 | |
| AK012220.1 | - | 1 - 2 | 150 | 71 | ||
| AK017685.1 | - | 1 - 2 | 161 | 71 | ||
| BC049574.1 | - | 1 - 2 | 356 | 6593108 | 71 | |
| BC052700.1 | - | 1 - 2 | 142 | 6416835 | 71 | |
| BC063057.1 | - | 1 - 2 | 135 | 5715784 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lix1
Genomic Map