Assay Details
| Gene Symbol: Cd209b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026972.5 | NP_081248.4 | 2 - 3 | 186 | 85 | |
| NR_104430.2 | 2 - 3 | 186 | 85 | |||
| GenBank mRNA | AF373409.1 | - | 2 - 3 | 164 | 85 | |
| AF422108.1 | - | 2 - 3 | 159 | 85 | ||
| AF424790.1 | - | 2 - 3 | 158 | 85 | ||
| AF424791.1 | - | 2 - 3 | 158 | 85 | ||
| AF424792.1 | - | 2 - 3 | 158 | 85 | ||
| AF424793.1 | - | 2 - 3 | 158 | 85 | ||
| AF424794.1 | - | 2 - 3 | 178 | 85 | ||
| AF424795.1 | - | 2 - 3 | 175 | 85 | ||
| AF424796.1 | - | 2 - 3 | 169 | 85 | ||
| AF424803.1 | - | 2 - 3 | 194 | 85 | ||
| AK007656.1 | - | 1 - 2 | 102 | 85 | ||
| AK138999.1 | - | 2 - 3 | 160 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd209b
Genomic Map