Assay Details
| Gene Symbol: Prpf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159714.1 | NP_001153186.1 | 2 - 3 | 263 | 83 | |
| NM_027328.4 | NP_081604.3 | 2 - 3 | 263 | 83 | ||
| GenBank mRNA | AK005294.1 | - | 2 - 3 | 231 | 83 | |
| AK012987.1 | - | 2 - 3 | 257 | 83 | ||
| AK033190.1 | - | 2 - 3 | 266 | 83 | ||
| AK033283.1 | - | 2 - 3 | 228 | 83 | ||
| AK051260.1 | - | 2 - 3 | 223 | 83 | ||
| AY040823.1 | - | 2 - 3 | 239 | 83 | ||
| BB839183.1 | - | 2 - 3 | 264 | 83 | ||
| BC018376.1 | - | 2 - 3 | 229 | 3673893 | 83 | |
| BC057877.1 | - | 1 - 2 | 206 | 5347609 | 83 | |
| BC061461.1 | - | 2 - 3 | 207 | 5717606 | 83 | |
| CT010189.1 | - | 1 - 2 | 183 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf31
Genomic Map