Assay Details
| Gene Symbol: Prpf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159714.1 | NP_001153186.1 | 6 - 7 | 608 | 64 | |
| NM_027328.4 | NP_081604.3 | 6 - 7 | 608 | 64 | ||
| GenBank mRNA | AK005294.1 | - | 6 - 7 | 576 | 64 | |
| AK044457.1 | - | 6 - 7 | 671 | 64 | ||
| AK051260.1 | - | 6 - 7 | 568 | 64 | ||
| AY040823.1 | - | 6 - 7 | 584 | 64 | ||
| BC018376.1 | - | 6 - 7 | 574 | 3673893 | 64 | |
| BC057877.1 | - | 5 - 6 | 551 | 5347609 | 64 | |
| BC061461.1 | - | 6 - 7 | 552 | 5717606 | 64 | |
| CT010189.1 | - | 5 - 6 | 528 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf31
Genomic Map