Assay Details
| Gene Symbol: Prpf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159714.1 | NP_001153186.1 | 8 - 9 | 935 | 111 | |
| NM_027328.4 | NP_081604.3 | 8 - 9 | 935 | 111 | ||
| GenBank mRNA | AK005294.1 | - | 8 - 9 | 903 | 111 | |
| AK033190.1 | - | 8 - 9 | 938 | 111 | ||
| AK033283.1 | - | 8 - 9 | 900 | 111 | ||
| AK044457.1 | - | 7 - 8 | 2336 | 111 | ||
| AK051260.1 | - | 8 - 9 | 895 | 111 | ||
| AY040823.1 | - | 8 - 9 | 911 | 111 | ||
| BC018376.1 | - | 8 - 9 | 901 | 3673893 | 111 | |
| BC057877.1 | - | 7 - 8 | 878 | 5347609 | 111 | |
| BC061461.1 | - | 8 - 9 | 879 | 5717606 | 111 | |
| CT010189.1 | - | 7 - 8 | 855 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf31
Genomic Map