Assay Details
| Gene Symbol: Prpf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159714.1 | NP_001153186.1 | 12 - 13 | 1338 | 58 | |
| NM_027328.4 | NP_081604.3 | 12 - 13 | 1356 | 58 | ||
| GenBank mRNA | AK005294.1 | - | 12 - 13 | 1323 | 58 | |
| AK033190.1 | - | 12 - 13 | 1359 | 58 | ||
| AK033283.1 | - | 12 - 13 | 1321 | 58 | ||
| AK051260.1 | - | 12 - 13 | 1316 | 58 | ||
| AY040823.1 | - | 12 - 13 | 1332 | 58 | ||
| BC018376.1 | - | 12 - 13 | 1322 | 3673893 | 58 | |
| BC057877.1 | - | 11 - 12 | 1299 | 5347609 | 58 | |
| BC061461.1 | - | 12 - 13 | 1282 | 5717606 | 58 | |
| CT010189.1 | - | 11 - 12 | 1276 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf31
Genomic Map