Assay Details
| Gene Symbol: Rcor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054048.3 | NP_473389.1 | 3 - 4 | 198 | 68 | |
| GenBank mRNA | AK052295.1 | - | 5 - 6 | 452 | 68 | |
| BC055719.1 | - | 4 - 5 | 619 | 6415591 | 68 | |
| X83587.2 | - | 3 - 4 | 200 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rcor2
Genomic Map