Assay Details
| Gene Symbol: Rcor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054048.3 | NP_473389.1 | 6 - 7 | 553 | 85 | |
| GenBank mRNA | AK052295.1 | - | 8 - 9 | 807 | 85 | |
| BC055719.1 | - | 7 - 8 | 974 | 6415591 | 85 | |
| X83587.2 | - | 6 - 7 | 555 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rcor2
Genomic Map