Assay Details
| Gene Symbol: Rcor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054048.3 | NP_473389.1 | 10 - 11 | 1136 | 63 | |
| GenBank mRNA | AK052295.1 | - | 12 - 13 | 1390 | 63 | |
| BC055719.1 | - | 11 - 12 | 1557 | 6415591 | 63 | |
| X83587.2 | - | 10 - 11 | 1138 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription, DNA-templated
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
enzyme binding
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription, DNA-templated
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
enzyme binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938930 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938926 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938928 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938929 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118784 | BC055719.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118784 | X83587.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118784 | NM_054048.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118784 | AK052295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118784 | AK205817.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824861 | AK054426.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824861 | AY355342.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824861 | BC066795.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824861 | NM_172481.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483921 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124395 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114831 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118784 | BC055719.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118784 | X83587.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118784 | NM_054048.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118784 | AK052295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118784 | AK205817.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P306689 | BC055719.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P306689 | X83587.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P306689 | NM_054048.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P306689 | AK052295.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P306689 | AK205817.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31938883 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114831 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03546009 | - | - |
| Whole Mouse Genome G4122A | A_51_P306689 | BC055719.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P306689 | X83587.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P306689 | NM_054048.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P306689 | AK052295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P306689 | AK205817.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105984 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114831 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14190927 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14190933 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14190930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14419381 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938930 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938929 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938926 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31938928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31939052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31938927 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14190924 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14257113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14436666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14301707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14273091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938932 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14301706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14300753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938923 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938870 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14301689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14273090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938868 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14412832 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938873 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14190922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938892 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14314893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938963 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14442497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938949 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14393137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938902 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14436667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14393124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938880 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938931 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938866 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938863 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14273087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938937 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31938971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14400730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14451021 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10461065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461070 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461063 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461069 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461066 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417302_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461057 | - | - |
| Mouse Genome 430 2.0 Array | 1417302_at | - | - |
| Mouse Genome 430 2.0 Array | 1417302_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 104383_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 104383_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900001773.mm.1 | NM_054048.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001773.mm.1 | TC1900000134.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417302_PM_at | - | - |
| Mouse Expression Set 430 | 1417302_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417302_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417302_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417302_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417302_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 696811 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5360717 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005360717 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1214713 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214713 | - | - |
| mouse-6 v1.1 (Illumina) | scl53547.13_444-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5360717 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53547.13_444-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360717 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1214713 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214713 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl53547.13_444-S | - | - |

Rcor2
Genomic Map