Assay Details
| Gene Symbol: Asb15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080847.3 | NP_543123.2 | 5 - 6 | 582 | 87 | |
| GenBank mRNA | AF403043.1 | - | 5 - 6 | 583 | 87 | |
| AK019556.1 | - | 5 - 6 | 583 | 87 | ||
| AK029759.1 | - | 5 - 6 | 514 | 87 | ||
| BC050794.1 | - | 5 - 6 | 531 | 6742480 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asb15
Genomic Map