Assay Details
| Gene Symbol: Pask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080850.2 | NP_543126.2 | 3 - 4 | 732 | 67 | |
| GenBank mRNA | AK028435.1 | - | 3 - 4 | 734 | 67 | |
| AK083528.1 | - | 3 - 4 | 703 | 67 | ||
| AK083590.1 | - | 3 - 4 | 697 | 67 | ||
| AK129063.1 | - | 3 - 4 | 622 | 67 | ||
| AK172031.1 | - | 3 - 4 | 680 | 67 | ||
| BC150821.1 | - | 2 - 3 | 444 | 9087732 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pask
Genomic Map