Assay Details
| Gene Symbol: Pask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080850.2 | NP_543126.2 | 9 - 10 | 1769 | 99 | |
| GenBank mRNA | AK028435.1 | - | 9 - 10 | 1771 | 99 | |
| AK083528.1 | - | 9 - 10 | 1740 | 99 | ||
| AK083590.1 | - | 9 - 10 | 1734 | 99 | ||
| AK129063.1 | - | 9 - 10 | 1659 | 99 | ||
| AK172031.1 | - | 9 - 10 | 1717 | 99 | ||
| BC150821.1 | - | 8 - 9 | 1481 | 9087732 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pask
Genomic Map