Assay Details
| Gene Symbol: Pask | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080850.2 | NP_543126.2 | 15 - 16 | 4025 | 99 | |
| GenBank mRNA | AK028435.1 | - | 15 - 16 | 4027 | 99 | |
| AK081074.1 | - | 728 | 99 | |||
| AK083528.1 | - | 15 - 16 | 3997 | 99 | ||
| AK083590.1 | - | 15 - 16 | 3991 | 99 | ||
| AK129063.1 | - | 15 - 16 | 3915 | 99 | ||
| AK141220.1 | - | 662 | 99 | |||
| BC150821.1 | - | 14 - 15 | 3737 | 9087732 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pask
Genomic Map