Assay Details
| Gene Symbol: Bcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024468.3 | NP_001019639.1 | 4 - 5 | 632 | 89 | |
| NM_007532.5 | NP_031558.3 | 4 - 5 | 651 | 89 | ||
| GenBank mRNA | AK012821.1 | - | 4 - 5 | 427 | 89 | |
| AK013888.1 | - | 4 - 5 | 588 | 89 | ||
| AK036309.1 | - | 4 - 5 | 633 | 89 | ||
| AK138468.1 | - | 4 - 5 | 545 | 89 | ||
| AK166971.1 | - | 4 - 5 | 587 | 89 | ||
| AK167232.1 | - | 4 - 5 | 578 | 89 | ||
| AK167368.1 | - | 4 - 5 | 497 | 89 | ||
| AK168249.1 | - | 4 - 5 | 681 | 89 | ||
| BC053706.1 | - | 4 - 5 | 583 | 30063465 | 89 | |
| U42443.1 | - | 3 - 4 | 388 | 89 | ||
| X17502.1 | - | 4 - 5 | 575 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bcat1
Genomic Map