Assay Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301250.1 | NP_001288179.1 | 5 - 6 | 803 | 59 | |
| NM_007602.4 | NP_031628.1 | 5 - 6 | 967 | 59 | ||
| GenBank mRNA | AK155179.1 | - | 5 - 6 | 945 | 59 | |
| AK164293.1 | - | 5 - 6 | 877 | 59 | ||
| AK170034.1 | - | 5 - 6 | 904 | 59 | ||
| BC014767.1 | - | 5 - 6 | 795 | 4458488 | 59 | |
| U85020.1 | - | 5 - 6 | 886 | 59 | ||
| Y10656.1 | - | 5 - 6 | 899 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn5
Genomic Map