Assay Details
| Gene Symbol: Capn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007603.3 | NP_031629.3 | 11 - 12 | 1825 | 75 | |
| GenBank mRNA | AK145116.1 | - | 11 - 12 | 1840 | 75 | |
| BC052167.1 | - | 11 - 12 | 1726 | 6517565 | 75 | |
| Y12582.1 | - | 11 - 12 | 1734 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn6
Genomic Map