Assay Details
| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008485.3 | NP_032511.3 | 4 - 5 | 713 | 68 | |
| GenBank mRNA | BB617855.1 | - | 3 - 4 | 477 | 68 | |
| U43327.2 | - | 3 - 4 | 446 | 68 | ||
More Information
Set Membership: |
|

Lamc2
Genomic Map