Assay Details
| Gene Symbol: Lamc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008485.3 | NP_032511.3 | 8 - 9 | 1264 | 85 | |
| GenBank mRNA | L20477.1 | - | 6 - 7 | 824 | 85 | |
| U43327.2 | - | 7 - 8 | 997 | 85 | ||
More Information
Set Membership: |
|

Lamc2
Genomic Map