Assay Details
| Gene Symbol: Ncoa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008679.3 | NP_032705.2 | 5 - 6 | 574 | 79 | |
| GenBank mRNA | AF000581.1 | - | 4 - 5 | 463 | 79 | |
| AK137507.1 | - | 5 - 6 | 560 | 79 | ||
| AK142306.1 | - | 5 - 6 | 479 | 79 | ||
| BC086482.1 | - | 5 - 6 | 544 | 30104508 | 79 | |
| BC093478.1 | - | 2 - 3 | 205 | 79 | ||
| BC141177.1 | - | 4 - 5 | 405 | 9055789 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncoa3
Genomic Map