Assay Details
| Gene Symbol: Pde9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163748.1 | NP_001157220.1 | 3 - 4 | 269 | 88 | |
| NM_008804.4 | NP_032830.3 | 3 - 4 | 269 | 88 | ||
| GenBank mRNA | AF031147.1 | - | 3 - 4 | 224 | 88 | |
| AF068247.1 | - | 3 - 4 | 233 | 88 | ||
| AK030509.1 | - | 3 - 4 | 253 | 88 | ||
| AK168143.1 | - | 3 - 4 | 251 | 88 | ||
| BC025882.1 | - | 3 - 4 | 227 | 88 | ||
| BC038675.1 | - | 3 - 4 | 218 | 88 | ||
| BC046915.1 | - | 3 - 4 | 218 | 88 | ||
| BC061163.1 | - | 3 - 4 | 249 | 6533665 | 88 | |
| BY283127.1 | - | 3 - 4 | 270 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde9a
Genomic Map