Assay Details
| Gene Symbol: Klk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008940.2 | NP_032966.1 | 2 - 3 | 721 | 118 | |
| GenBank mRNA | BC055895.1 | - | 2 - 3 | 235 | 4019947 | 118 |
| D30785.1 | - | 2 - 3 | 721 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klk8
Genomic Map