Assay Details
| Gene Symbol: Ptpn14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008976.2 | NP_033002.2 | 4 - 5 | 978 | 68 | |
| GenBank mRNA | AF170902.1 | - | 3 - 4 | 545 | 68 | |
| AF170903.1 | - | 3 - 4 | 545 | 68 | ||
| AK028783.1 | - | 4 - 5 | 942 | 68 | ||
| AK037646.1 | - | 4 - 5 | 979 | 68 | ||
| AK086524.1 | - | 4 - 5 | 972 | 68 | ||
| AK156480.1 | - | 4 - 5 | 724 | 68 | ||
| D31842.1 | - | 3 - 4 | 575 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn14
Genomic Map