Assay Details
| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127177.1 | NP_001120649.1 | 7 - 8 | 966 | 145 | |
| NM_008977.3 | NP_033003.1 | 7 - 8 | 966 | 145 | ||
| GenBank mRNA | AK014528.1 | - | 7 - 8 | 933 | 145 | |
| AK076072.1 | - | 7 - 8 | 919 | 145 | ||
| AK132013.1 | - | 7 - 8 | 929 | 145 | ||
| BC008269.1 | - | 7 - 8 | 940 | 3602983 | 145 | |
| M80739.1 | - | 7 - 8 | 921 | 145 | ||
| M81477.1 | - | 7 - 8 | 921 | 145 | ||
| S52655.1 | - | 7 - 8 | 916 | 145 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn2
Genomic Map