Assay Details
| Gene Symbol: Rgs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009062.3 | NP_033088.2 | 1 - 2 | 168 | 116 | |
| GenBank mRNA | AB004315.1 | - | 1 - 2 | 49 | 116 | |
| AK030536.1 | - | 1 - 2 | 169 | 116 | ||
| AK039134.1 | - | 1 - 2 | 169 | 116 | ||
| BC003882.1 | - | 1 - 2 | 146 | 3496887 | 116 | |
| BC055293.1 | - | 1 - 2 | 158 | 6494449 | 116 | |
| DQ346659.1 | - | 1 - 2 | 168 | 116 | ||
| DQ346660.1 | - | 1 - 2 | 168 | 116 | ||
| DQ346661.1 | - | 1 - 2 | 168 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Generation and characterization of rgs5 mutant mice.
- Authors
- Nisancioglu MH, Mahoney WM Jr, Kimmel DD, Schwartz SM, Betsholtz C, Genove G
- Journal
- Mol Cell Biol - Page No 2324 (Tue Apr 01 00:00:00 UTC 2008)
- Volume (Year)
- 28 (2008)
- Product Usage
- -

Rgs4
Genomic Map