Assay Details
| Gene Symbol: Coq7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF080580.1 | - | 1 - 2 | 67 | 95 | |
| AF098949.1 | - | 1 - 2 | 133 | 95 | ||
| AK137841.1 | - | 1 - 2 | 131 | 95 | ||
| AK158380.1 | - | 1 - 2 | 118 | 95 | ||
| BC038681.1 | - | 1 - 2 | 100 | 1380996 | 95 | |
| BY284216.1 | - | 1 - 2 | 172 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Coq7
Genomic Map