Assay Details
| Gene Symbol: Coq7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF053770.1 | - | 1 - 2 | 186 | 95 | |
| AF080580.1 | - | 2 - 3 | 249 | 95 | ||
| AF098949.1 | - | 2 - 3 | 315 | 95 | ||
| AK158380.1 | - | 2 - 3 | 300 | 95 | ||
| BC038681.1 | - | 2 - 3 | 282 | 1380996 | 95 | |
| U81277.1 | - | 1 - 2 | 31 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Coq7
Genomic Map