Assay Details
| Gene Symbol: P2rx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011026.2 | NP_035156.2 | 1 - 2 | 234 | 66 | |
| GenBank mRNA | AF089751.1 | - | 1 - 2 | 168 | 66 | |
| AF089752.1 | - | 1 - 2 | 168 | 66 | ||
| AJ251459.1 | - | 1 - 2 | 148 | 66 | ||
| AJ251460.1 | - | 1 - 2 | 148 | 66 | ||
| AJ251461.1 | - | 1 - 2 | 148 | 66 | ||
| AJ251462.1 | - | 1 - 2 | 148 | 66 | ||
| AK150351.1 | - | 1 - 2 | 224 | 66 | ||
| AK162820.1 | - | 1 - 2 | 200 | 66 | ||
| AK163104.1 | - | 1 - 2 | 222 | 66 | ||
| BC002099.1 | - | 1 - 2 | 207 | 66 | ||
| BC005597.1 | - | 1 - 2 | 182 | 3482458 | 66 | |
| BY150918.1 | - | 1 - 2 | 248 | 66 | ||
| JX417451.1 | - | 1 - 2 | 194 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P2rx4
Genomic Map