Assay Details
| Gene Symbol: P2rx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011026.2 | NP_035156.2 | 2 - 3 | 383 | 75 | |
| GenBank mRNA | AF089751.1 | - | 2 - 3 | 317 | 75 | |
| AF089752.1 | - | 2 - 3 | 317 | 75 | ||
| AJ251459.1 | - | 2 - 3 | 297 | 75 | ||
| AJ251460.1 | - | 2 - 3 | 297 | 75 | ||
| AJ251461.1 | - | 2 - 3 | 297 | 75 | ||
| AJ251462.1 | - | 2 - 3 | 297 | 75 | ||
| AK150351.1 | - | 2 - 3 | 373 | 75 | ||
| AK162820.1 | - | 2 - 3 | 349 | 75 | ||
| AK163104.1 | - | 2 - 3 | 371 | 75 | ||
| BC002099.1 | - | 2 - 3 | 356 | 75 | ||
| BC005597.1 | - | 2 - 3 | 331 | 3482458 | 75 | |
| JX417451.1 | - | 2 - 3 | 343 | 75 | ||
More Information
Set Membership: |
|

P2rx4
Genomic Map