Assay Details
| Gene Symbol: P2rx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF089752.1 | - | 5 - 6 | 562 | 74 | |
| AJ251460.1 | - | 5 - 6 | 542 | 74 | ||
| AJ251462.1 | - | 5 - 6 | 542 | 74 | ||
| BC005597.1 | - | 5 - 6 | 576 | 3482458 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

P2rx4
Genomic Map