Assay Details
| Gene Symbol: Med1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080118.1 | NP_001073587.1 | 10 - 11 | 948 | 67 | |
| NM_013634.2 | NP_038662.2 | 10 - 11 | 813 | 67 | ||
| NM_134027.2 | NP_598788.2 | 10 - 11 | 948 | 67 | ||
| GenBank mRNA | AF000294.1 | - | 10 - 11 | 813 | 67 | |
| AF332073.1 | - | 9 - 10 | 693 | 67 | ||
| AF332074.1 | - | 9 - 10 | 693 | 67 | ||
| AK049203.1 | - | 10 - 11 | 939 | 67 | ||
| AK054437.1 | - | 10 - 11 | 949 | 67 | ||
| AK147199.1 | - | 10 - 11 | 925 | 67 | ||
| BC021440.1 | - | 10 - 11 | 919 | 67 | ||
| BC079636.1 | - | 10 - 11 | 918 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Med1
Genomic Map