Assay Details
| Gene Symbol: Zfp276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020497.2 | NP_065243.2 | 7 - 8 | 1800 | 75 | |
| GenBank mRNA | AF178935.1 | - | 6 - 7 | 842 | 75 | |
| AK028939.1 | - | 7 - 8 | 1753 | 75 | ||
| AK133363.1 | - | 7 - 8 | 1801 | 75 | ||
| BC078415.1 | - | 7 - 8 | 1184 | 75 | ||
| BC138579.1 | - | 6 - 7 | 792 | 8861599 | 75 | |
| BC138580.1 | - | 6 - 7 | 792 | 8861600 | 75 | |
| BC145340.1 | - | 6 - 7 | 792 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp276
Genomic Map