Assay Details
| Gene Symbol: Piwil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021308.1 | NP_067283.1 | 5 - 6 | 827 | 77 | |
| GenBank mRNA | AB032605.1 | - | 5 - 6 | 827 | 77 | |
| AK030116.1 | - | 5 - 6 | 771 | 77 | ||
| BC138444.1 | - | 5 - 6 | 815 | 8861464 | 77 | |
| BC145717.1 | - | 5 - 6 | 815 | 40131020 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Piwil2
Genomic Map