Assay Details
| Gene Symbol: Myg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021713.2 | NP_068359.1 | 1 - 2 | 243 | 62 | |
| GenBank mRNA | AF289484.1 | - | 1 - 2 | 235 | 62 | |
| AK002656.1 | - | 1 - 2 | 218 | 62 | ||
| AK004104.1 | - | 1 - 2 | 218 | 62 | ||
| AK013232.1 | - | 1 - 2 | 189 | 62 | ||
| BC028501.1 | - | 1 - 2 | 228 | 5256915 | 62 | |
| CJ180315.1 | - | 1 - 2 | 243 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myg1
Genomic Map