Assay Details
| Gene Symbol: Wdr54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023790.2 | NP_076279.1 | 4 - 5 | 553 | 61 | |
| GenBank mRNA | AF053628.1 | - | 4 - 5 | 541 | 61 | |
| AK006536.1 | - | 4 - 5 | 453 | 61 | ||
| AK154345.1 | - | 4 - 5 | 544 | 61 | ||
| AK202399.1 | - | 3 - 4 | 168 | 61 | ||
| BC083332.1 | - | 5 - 6 | 458 | 6819838 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr54
Genomic Map