Assay Details
| Gene Symbol: Slc38a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199217.1 | NP_001186146.1 | 8 - 9 | 772 | 75 | |
| NM_001199218.1 | NP_001186147.1 | 8 - 9 | 792 | 75 | ||
| NM_023805.3 | NP_076294.2 | 8 - 9 | 940 | 75 | ||
| GenBank mRNA | AF159856.1 | - | 8 - 9 | 742 | 75 | |
| AK002607.1 | - | 8 - 9 | 758 | 75 | ||
| AK035155.1 | - | 960 | 75 | |||
| BC010678.1 | - | 8 - 9 | 772 | 75 | ||
| BC021324.1 | - | 8 - 9 | 826 | 75 | ||
| BC054846.1 | - | 8 - 9 | 915 | 6493334 | 75 | |
| BC055339.1 | - | 8 - 9 | 792 | 6415115 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a3
Genomic Map