Assay Details
| Gene Symbol: Tril | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025817.4 | NP_080093.1 | 1 | 1881 | 62 | |
| GenBank mRNA | AK004681.2 | - | 1 | 1552 | 62 | |
| AK033401.1 | - | 1 | 1544 | 62 | ||
| AK051421.1 | - | 1 | 1081 | 62 | ||
| AK122338.1 | - | 1 | 1469 | 62 | ||
| BC043099.1 | - | 1 | 1750 | 6406165 | 62 | |
| BC059224.1 | - | 1 | 1750 | 6406165 | 62 | |
| Gene Symbol: Gm16499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039090.1 | - | 62 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tril
Genomic Map