Assay Details
| Gene Symbol: Tlcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291235.1 | NP_001278164.1 | 2 - 3 | 166 | 75 | |
| NM_001291236.1 | NP_001278165.1 | 2 - 3 | 642 | 75 | ||
| NM_001291237.1 | NP_001278166.1 | 3 - 4 | 350 | 75 | ||
| NM_026708.2 | NP_080984.1 | 2 - 3 | 477 | 75 | ||
| GenBank mRNA | AA671983.1 | - | 3 - 4 | 367 | 75 | |
| AK002265.1 | - | 2 - 3 | 425 | 75 | ||
| AK002703.1 | - | 2 - 3 | 373 | 75 | ||
| AK046577.1 | - | 3 - 4 | 2357 | 75 | ||
| AK161721.1 | - | 377 | 75 | |||
| AK165432.1 | - | 589 | 75 | |||
| BC005702.1 | - | 2 - 3 | 369 | 3598444 | 75 | |
| BC039182.1 | - | 2 - 3 | 612 | 3500908 | 75 | |
| BG083251.2 | - | 2 - 3 | 224 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tlcd1
Genomic Map