Assay Details
| Gene Symbol: Hddc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026812.2 | NP_081088.1 | 1 - 2 | 112 | 81 | |
| GenBank mRNA | AK004078.1 | - | 1 - 2 | 113 | 81 | |
| BC038310.1 | - | 1 - 2 | 125 | 5119020 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25266151 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521882 | BC055847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521882 | BC038310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521882 | NM_026812.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521882 | AK004078.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521882 | BC055847.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521882 | BC038310.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521882 | NM_026812.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P521882 | AK004078.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47817 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137237 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521882 | BC055847.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521882 | BC038310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521882 | NM_026812.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521882 | AK004078.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521882 | BC055847.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521882 | BC038310.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521882 | NM_026812.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P521882 | AK004078.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25266157 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137237 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01558595 | - | - |
| Whole Mouse Genome G4122A | A_52_P521882 | BC055847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521882 | BC038310.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521882 | NM_026812.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521882 | AK004078.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521882 | BC055847.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521882 | BC038310.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521882 | NM_026812.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P521882 | AK004078.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104990 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137237 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06134786 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25266151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25266147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05887075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06221000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06444680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06341707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06106112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06341710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06220994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06584600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06444684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06341706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06603414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06106113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06603410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06022405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25266145 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10554466 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554465 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554467 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452875_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428692_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10554463 | - | - |
| Mouse Genome 430 2.0 Array | 1428692_at | - | - |
| Mouse Genome 430 2.0 Array | 1452875_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428692_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452875_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68695_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68695_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700010176.mm.1 | NM_026812.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010176.mm.1 | NM_026812.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010176.mm.1 | TC0700001152.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010175.mm.1 | NM_026812.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010175.mm.1 | NM_026812.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010175.mm.1 | TC0700001152.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010177.mm.1 | NM_026812.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010177.mm.1 | NM_026812.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010177.mm.1 | TC0700001152.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700010174.mm.1 | NM_026812.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010174.mm.1 | NM_026812.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700010174.mm.1 | TC0700001152.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428692_at | - | - |
| Mouse Expression Set 430 | 1452875_PM_at | - | - |
| Mouse Expression Set 430 | 1428692_PM_at | - | - |
| Mouse Expression Set 430 | 1452875_at | - | - |
| Murine Genome U74v2 Set | 130319_at | - | - |
| Murine Genome U74v2 Set | 130319_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428692_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452875_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452875_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428692_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 636286 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4230017 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104200170 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4230017 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004230017 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244961 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248948 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244961 | - | - |
| mouse-6 v1.1 (Illumina) | scl0068695.1_200-S | - | - |
| mouse-6 v1.1 (Illumina) | scl31128.1.1_88-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4230017 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104200170 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0068695.1_200-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4230017 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104200170 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1248948 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248948 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0068695.1_200-S | - | - |

Hddc3
Genomic Map