Assay Details
| Gene Symbol: Srpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083895.3 | NP_001077364.2 | 3 - 4 | 661 | 135 | |
| NM_026838.4 | NP_081114.2 | 4 - 5 | 713 | 135 | ||
| GenBank mRNA | AK136935.1 | - | 3 - 4 | 516 | 135 | |
| BC028307.1 | - | 4 - 5 | 442 | 3488526 | 135 | |
| CN539245.1 | - | 4 - 5 | 476 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Srpx2
Genomic Map