Assay Details
| Gene Symbol: Srpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083895.3 | NP_001077364.2 | 8 - 9 | 1448 | 71 | |
| NM_026838.4 | NP_081114.2 | 9 - 10 | 1500 | 71 | ||
| GenBank mRNA | AF393640.1 | - | 7 - 8 | 1018 | 71 | |
| AK136935.1 | - | 8 - 9 | 1303 | 71 | ||
| BC028307.1 | - | 9 - 10 | 1229 | 3488526 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Srpx2
Genomic Map