Assay Details
| Gene Symbol: Arv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026855.4 | NP_081131.1 | 4 - 5 | 107 | ||
| GenBank mRNA | AK004398.1 | - | 4 - 5 | 107 | ||
| AK042561.1 | - | 2 - 3 | 107 | |||
| BC119376.1 | - | 4 - 5 | 8734125 | 107 | ||
| BC120631.1 | - | 4 - 5 | 40129554 | 107 | ||
| BC144875.1 | - | 4 - 5 | 107 | |||
| BC144876.1 | - | 4 - 5 | 107 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Arv1
Genomic Map