Assay Details
| Gene Symbol: Mvb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028617.2 | NP_082893.1 | 3 - 4 | 326 | 68 | |
| GenBank mRNA | AK003644.1 | - | 3 - 4 | 306 | 68 | |
| BC020119.1 | - | 3 - 4 | 326 | 3982577 | 68 | |
| BC027419.1 | - | 3 - 4 | 302 | 4163554 | 68 | |
| BC087734.1 | - | 3 - 4 | 323 | 5101734 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25957393 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25957392 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146303 | AK003644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146303 | BC020119.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146303 | NM_028617.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146303 | AK003644.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146303 | BC020119.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P146303 | NM_028617.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211755 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760404 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P129371 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146303 | AK003644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146303 | BC020119.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146303 | NM_028617.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146303 | AK003644.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146303 | BC020119.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P146303 | NM_028617.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25957392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25957396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25957395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25957403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25957400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25957405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25957398 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P129371 | - | - |
| Whole Mouse Genome G4122A | A_51_P146303 | AK003644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146303 | BC020119.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146303 | NM_028617.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146303 | AK003644.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146303 | BC020119.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P146303 | NM_028617.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100688 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P129371 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06871166 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25957393 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25957392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25957407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07407820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07407817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06778087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06773584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06871163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07439014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07260420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07316099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06856851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06672817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06773585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06713141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25957389 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10572639 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572640 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572638 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572645 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572646 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572641 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424773_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437043_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455895_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572637 | - | - |
| Mouse Genome 430 2.0 Array | 1424773_at | - | - |
| Mouse Genome 430 2.0 Array | 1455895_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1424773_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455895_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1437043_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1437043_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73711_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73711_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800005736.mm.1 | NM_028617.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005736.mm.1 | NM_028617.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800005736.mm.1 | TC0800000795.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455895_PM_x_at | - | - |
| Mouse Expression Set 430 | 1424773_PM_at | - | - |
| Mouse Expression Set 430 | 1424773_at | - | - |
| Mouse Expression Set 430 | 1437043_PM_a_at | - | - |
| Mouse Expression Set 430 | 1455895_x_at | - | - |
| Mouse Expression Set 430 | 1437043_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437043_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424773_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424773_at | - | - |
| Mouse Genome 430A 2.0 Array | 1437043_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455895_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455895_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437043_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424773_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455895_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455895_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424773_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1437043_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 466093 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1400673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001400673 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2753109 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2753109 | - | - |
| mouse-6 v1.1 (Illumina) | scl33697.10.1_67-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1400673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33697.10.1_67-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1400673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2753109 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2753109 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33697.10.1_67-S | - | - |

Mvb12a
Genomic Map