Assay Details
| Gene Symbol: Tjap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028751.3 | NP_083027.1 | 3 - 4 | 289 | 59 | |
| GenBank mRNA | AK002882.2 | - | 3 - 4 | 276 | 59 | |
| AK188857.1 | - | 1 - 2 | 125 | 59 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK002882.2,NM_028751.3 transcripts.
Set Membership: |
|

Tjap1
Genomic Map