Assay Details
| Gene Symbol: Hvcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042489.2 | NP_001035954.1 | 5 - 6 | 753 | 57 | |
| NM_028752.3 | NP_083028.1 | 4 - 5 | 615 | 57 | ||
| GenBank mRNA | AK002854.1 | - | 4 - 5 | 616 | 57 | |
| AK154880.1 | - | 5 - 6 | 645 | 57 | ||
| AK155123.1 | - | 5 - 6 | 590 | 57 | ||
| AK170422.1 | - | 5 - 6 | 614 | 57 | ||
| BC021548.1 | - | 5 - 6 | 584 | 4913027 | 57 | |
| BY703352.1 | - | 4 - 5 | 609 | 57 | ||
| CF734112.1 | - | 5 - 6 | 588 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hvcn1
Genomic Map