Assay Details
| Gene Symbol: Lmbr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029098.3 | NP_083374.1 | 8 - 9 | 870 | 61 | |
| GenBank mRNA | AK003656.1 | - | 8 - 9 | 802 | 61 | |
| AK030589.1 | - | 8 - 9 | 836 | 61 | ||
| AK049110.1 | - | 8 - 9 | 871 | 61 | ||
| AK143891.1 | - | 8 - 9 | 831 | 61 | ||
| AK154968.1 | - | 8 - 9 | 1004 | 61 | ||
| AK170328.1 | - | 8 - 9 | 831 | 61 | ||
| AK173121.1 | - | 8 - 9 | 1251 | 61 | ||
| AY052398.1 | - | 8 - 9 | 713 | 61 | ||
| BC023397.1 | - | 8 - 9 | 807 | 5043877 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbr1l
Genomic Map