Assay Details
| Gene Symbol: Nt5c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164363.1 | NP_001157835.1 | 4 - 5 | 803 | 88 | |
| NM_001164365.1 | NP_001157837.1 | 4 - 5 | 460 | 88 | ||
| NM_029810.4 | NP_084086.3 | 4 - 5 | 391 | 88 | ||
| NR_028353.1 | 5 - 6 | 450 | 88 | |||
| GenBank mRNA | AK008045.1 | - | 4 - 5 | 338 | 88 | |
| AK030786.1 | - | 5 - 6 | 408 | 88 | ||
| AK036730.1 | - | 4 - 5 | 349 | 88 | ||
| AK038458.1 | - | 1 - 2 | 96 | 88 | ||
| AK147901.1 | - | 4 - 5 | 349 | 88 | ||
| AK169616.1 | - | 4 - 5 | 349 | 88 | ||
| AK181552.1 | - | 3 - 4 | 252 | 88 | ||
| AK182698.1 | - | 2 - 3 | 136 | 88 | ||
| AK187621.1 | - | 3 - 4 | 252 | 88 | ||
| AK201286.1 | - | 2 - 3 | 193 | 88 | ||
| AK211414.1 | - | 2 - 3 | 193 | 88 | ||
| BC064760.1 | - | 3 - 4 | 325 | 30436992 | 88 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC006028.1 | - | Chr. - - [-] on Build | 88 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nt5c2
Genomic Map